54020-U
ORBO™ 555 LpDNPH 300/150 mg Sorbent Tube
W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 20 ea
Synonym(s):
LpDNPH ORBO™ Tube
About This Item
Recommended Products
material
W,W,W separators
Quality Level
Agency
NIOSH 2016 (Appendix B)
product line
ORBO™
composition
Bed A, 300 mg
Bed B, 150 mg
packaging
pkg of 20 ea
manufacturer/tradename
ORBO™ 555
technique(s)
active air sampling: suitable
O.D. × L
6 mm × 105 mm
matrix
2,4-dinitrophenylhydrazine (DNPH) on High Purity Silica Gel
particle size
20-40 mesh
application(s)
air monitoring
environmental
industrial hygiene
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.
Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.
Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.
Air monitoring is used to measure the level of air pollutants in environmental, industrial, or indoor settings to assess the air quality per standards established by health and regulatory agencies.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service