24442
SGE Needles for RN syringes
volume 10 μL, needle size 23 ga, needle L × O.D. 42 mm × 0.63 mm
Synonym(s):
REPL.NEEDLE N10-S-0.63, PK2
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
description
10R-S-0.63
feature
needle cone tip
packaging
pkg of 2 ea
manufacturer/tradename
SGE 037747
needle L × O.D.
42 mm × 0.63 mm
volume
10 μL
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service