Skip to Content
Merck
All Photos(1)

Documents

20042

Supelco

ORBO 351 4-TBC on Activated Coconut Charcoal (20/40), 100/50 mg

W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea

Synonym(s):

TBC on Charcoal Tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47131900

material

W,W,W separators

Quality Level

Agency

OSHA 56

product line

ORBO

composition

Bed A, 100 mg
Bed B, 50 mg

packaging

pkg of 50 ea

manufacturer/tradename

ORBO 351

technique(s)

active air sampling: suitable

O.D. × L

6 mm × 75 mm

matrix

4-tert-butyl catechol on charcoal
charcoal

matrix active group

4-tert-butylcatechol

application(s)

air monitoring
environmental
industrial hygiene

compatibility

mode of use Solvent desorption
technique used for Air monitoring

Looking for similar products? Visit Product Comparison Guide

General description

  • ORBO 351 4-TBC on Activated Coconut Charcoal (20/40), 100/50 mg solvent desorption tube is designed for gas and vapor sampling.
  • It contains two beds of the same selective adsorbent separated by glass wool or foam.
  • Dual-layer or two-bed construction of the tube allows for any sample breakthrough to be captured in the smaller back-up bed.

Legal Information

ORBO is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service