Skip to Content
Merck
All Photos(10)

Key Documents

ZRB2181

Sigma-Aldrich

Anti-KLF2 Antibody, clone 1C3 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-Nephrin Antibody, clone 1D16 ZooMAb® Rabbit Monoclonal, Krueppel-like factor 2, Lung krueppel-like factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C3, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 37.42 kDa
observed mol wt ~40 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... KLF2(10365)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C3 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects KLF2. It targets an epitope within 20 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human KLF2.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell nuclear extract.

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected KLF2 in HeLa cell nuclear extract.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution of this antibody detected KLF2 in Human stomach tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected KLF2 in MCF-7 cells.

Flow Cytometry Analysis: 0.1 g from a representative lot detected KLF2 in one million MCF-7 cells.

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected GST-tagged recombinant fragment corresponding to 56 amino acids from the N-terminal region of Human KLF2.

Affinity Binding Assay: A representative lot of this antibody bound KLF2 peptide with a KD of 9.2 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Krueppel-like factor 2 (UniProt: Q9Y5W3; also known as Lung krueppel-like factor, KLF2) is encoded by the KLF2 (also known as LKLF) gene (Gene ID: 10365) in human. KLF2 is a member of the krueppel C2H2-type zinc-finger protein family that serves as a transcription factor that binds to the CACCC box in the promoter of target genes and activates their transcription. It contains a nine amino acids transactivation domain (aa 43-51). Its expression is observed in mammalian development and is found in many different cell types. It plays a role in several biological processes including adipogenesis, embryonic erythropoiesis, epithelial integrity, inflammation, and T-cell viability. It is reported to inhibit proinflammatory activation of monocytes and KLF2 expression in monocytes is reduced by cytokine activation. Its expression in circulating monocytes is significantly reduced in subjects with chronic inflammatory conditions. It is shown to inhibit the transcriptional activity of both NF- B a

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service