The sequence information provided in the product description (epitope is within 23 amino acids from the N-terminal half) is in reference to the mature form of IL-18.
ZRB2106
Anti-IL-18 Antibody, clone 1N23 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
Synonym(s):
Interleukin-18;+C8Iboctadekin,;Interferon gamma-inducing factor;+C8IFN-gamma-inducing factor;Interleukin-1 gamma;IL-1 gamma
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1N23, recombinant monoclonal
description
1N23 Clone
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 22.33 kDa
observed mol wt ~22 kDa
purified by
using Protein A
species reactivity
human
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal half
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... IL18(3606)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in PC3 cell lysate.
Western Blotting Analysis: A 1:10,000 dilution of this antibody detected IL-18 in PC3 cell lysate.
Tested Applications
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected IL-18 in human tonsil tissue sections.
Affinity Binding Assay: A representative lot of this antibody bound IL-18 peptide with a KD of 1.2 x 10-8 in an affinity binding assay.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected IL-18 in THP-1 cells.
Flow Cytometry Analysis: 1 µg from a representative lot detected IL-18 in one million THP-1 cells.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Reconstitution
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
When you say that it recognizes an epitope within the first 23aa of the N-terminal half, are you talking about the N-terminal half of the mature form or of the pro-form of IL-18 (i.e., M1-L23 or Y37-D59?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service