Z756679
Freezer storage box, 81-place
assorted colors
Synonym(s):
freezer container, freezer storage, storage container
About This Item
Recommended Products
material
assorted colors
natural polypropylene
feature
Holds 81 x 0.5, 1.5 or 2 mL vials
packaging
pkg of 5 ea
manufacturer/tradename
Bel-Art F18852-0016
technique(s)
cryopreservation: suitable
L × W × H
14 cm × 14 cm × 4.9 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Alphanumeric indexing
- Withstand -80°C to +121°C (-112° to +249°F)
- Stackable with molded-in ′no slide′ features
- Lift off lid has matte panels on two sides for convenient labeling
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service