Z743501
Breathe-easier breathable tube membranes for microtubes
white, Rayon, pre-cut, (Sterile)
Synonym(s):
breathable sealing membrane, cell culture, microplates, microwell plates, multi-well plates, multi-well sealing film, plate sealers, plate sealing film, sealing film, tissue culture
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
Rayon
sterility
(Sterile)
feature
pre-cut
packaging
pack of 100 ea
manufacturer/tradename
Diversified Biotech BMTM-2000
color
white
General description
Breathe-EASIER (a white non-woven rayon membrane) transmits water vapor 6 times faster than Breathe-EASY for accelerated growth.
Membranes are ideal for cell growth, concentrating samples (e.g. freeze drying), and gas exchange.
Easy microtube cap closure. Will not bind in centrifuge rotors.
Formatted with an easy-to-use finger tab. Pre-cut to fit a variety of popular tube sizes. Convenient scoring makes liner removal quick and easy.
Membranes are gamma irradiated to ensure sterility. Free of cytotoxins.
Membranes are ideal for cell growth, concentrating samples (e.g. freeze drying), and gas exchange.
Easy microtube cap closure. Will not bind in centrifuge rotors.
Formatted with an easy-to-use finger tab. Pre-cut to fit a variety of popular tube sizes. Convenient scoring makes liner removal quick and easy.
Membranes are gamma irradiated to ensure sterility. Free of cytotoxins.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service