Key Documents
Z717304
Nichipet EX-Plus II pipette
volume 2-20 μL
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Pricing and availability is not currently available.
feature
electronic: no
single-channel
packaging
pack of 1 ea
pkg of 1 ea
manufacturer/tradename
Nichiryo 00-NPL02-20
availability
not available in (Not available in France)
volume
2-20 μL
General description
The Nichipet EX-Plus ll is a new digital micropipette addition to the Nichiryo family of pipettors. The Nichipet EX-Plus ll offers a new, key feature of one-touch calibration.
Time and money spent on the calibration of pipettes is now greatly reduced!
  • UV resistant
  • Solvent resistant
  • Fully autoclavable (121°C for 20 minutes)
  • One-touch calibration
  • Ceramic pistons
  • Filtered nozzles
  • Easy, convenient locking lever
  • Ergonomic design
  • Single channel, manual, continuously adjustable volume
  • Advanced durability against organic solvent dispensing
  • PTFE (Fluorine resin) in the air tight chamber of the instrument
  • A Perfluoro rubber O-ring with low cubical expansion in the airtight parts
  • Plunger made from a material resistant to organic solvents
  • An alloy spring to avoid metal corrosion by organic solvents
  • Quick and easy to use
Choose from one of the most recent versions:
Certificates of Analysis (COA)