Key Documents
Z353736
Sigma-Aldrich
Nalgene® centrifuge bottles, wide-mouth with sealing caps Style 3141
Nalgene® centrifuge bottles, wide-mouth with sealing caps Style 3141
1 of 1 reviewers received a sample product or took part in a promotion
nominal capacity 250 mL
Synonym(s):
leak proof centrifuge bottle
About This Item
UNSPSC Code:
41121703
NACRES:
NB.78
Pricing and availability is not currently available.
material
flat bottom bottle
polypropylene copolymer (with sealing closure)
translucent bottle
Joint
Joints threaded neck (58)
feature
RCF 27,500 × g
with cap
packaging
pack of 4 ea
manufacturer/tradename
Nalgene® 3141-0250
parameter
27500 × g max. RCF
nominal capacity
250 mL
General description
Centrifugalize low- to moderate-speed, large-volume, biological and chemical samples with these PPCO centrifuge bottles, which include a sealing closure for added leakproof assurance at maximum speeds when spinning hazardous materials. Molded of rigid and robust polypropylene copolymer, these translucent bottles are designed for pelleting applications, such as cell culture harvesting and protein biochemistry, including ammonium sulfate precipitations. Autoclavable with excellent mechanical strength.
Features and Benefits
  • Sturdy polypropylene copolymer (PPCO).
  • Designed for low- to moderate-centrifugation speed, large-volume, biological and chemical samples.
  • Ideal for use in cell culture harvesting and protein biochemistry applications, including ammonium sulfate precipitations.
  • Features a leakproof sealing closure for sample protection.
  • Autoclave safe.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–2 of 2 Questions  
  1. Is this product autoclavable? If so, at what parameters?
    1 answer
    1. Yes, this product is autoclavable. The recommended protocol is 121°C and 15 psig for 15 minutes.
      Helpful?
  2. Does the sealing cap have a built-in gasket, or an O-ring?
    1 answer
    1. This product features a simple friction closure in the form of a screw cap. A built-in gasket or O-ring is not included.
      Helpful?
Reviews
1 of 1 reviewers received a sample product or took part in a promotion
Active Filters
    • Review 1
    • Votes 0
    4 out of 5 stars.
    Associate Research Scientist
    This product work fine with high speed centrifuge and easy to clean
    Helpful?