Skip to Content
Merck
All Photos(5)

Key Documents

WH0057167M3

Sigma-Aldrich

Monoclonal Anti-SALL4 antibody produced in mouse

clone 6E3, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DRRS, Anti-HSAL4, Anti-MGC133050, Anti-dJ1112F19.1, Anti-sal-like 4 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6E3, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

ELISA: suitable
capture ELISA: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SALL4(57167)

General description

Spalt like transcription factor 4 (SALL4) is encoded by the gene with four exons, mapped to human chromosome 20q13.13-q13.2. The encoded protein belongs to the spalt-like protein family. SALL4 is characterized with a multiple zinc finger (ZnF) domains including one N-terminal C2HC-type ZnF and seven C2H2-type ZnF domains. In vertebrates, SALL4 is abundantly expressed in both embryonic and adult stem/stem-like cells.
The protein encoded by this gene may be a zinc finger transcription factor. Defects in this gene are a cause of Duane-radial ray syndrome (DRRS). (provided by RefSeq)

Immunogen

SALL4 (NP_065169, 954 a.a. ~ 1053 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
PKEILAPSVNVDPVVWNQYTSMLNGGLAVKTNEISVIQSGGVPTLPVSLGATSVVNNATVSKMDGSQSGISADVEKPSATDGVPKHQFPHFLEENKIAVS

Application

Monoclonal Anti-SALL4 antibody produced in mouse has been used in immunohistochemical staining.

Biochem/physiol Actions

Spalt like transcription factor 4 (SALL4) plays a crucial role in the regulation of cell stemness in biological development and tumor growth. Thus, this protein can be considered as a potent target for gene therapy. In addition, it also serves as a potential diagnostic marker for testicular germ cell tumors (GCTs). Polymorphisms in the gene are associated with the development of various diseases such as Duane-radial ray syndrome (DRRS, Okihiro syndrome), acro-renal-ocular syndrome (AROS), and SALL4-related Holt-Oram syndrome (HOS).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sall4 interacts with Nanog and co-occupies Nanog genomic sites in embryonic stem cells.
Wu Q, et al.
The Journal of Biological Chemistry, 24090-4 null
RNA-binding protein LIN28 is a marker for testicular germ cell tumors
Cao D, et al.
Human Pathology, 710-8 null
SALL4: engine of cell stemness
Xiong J.
Current gene therapy, 400-11 null
Multigene Deletions on Chromosome 20q13.13-q13.2 Including SALL4 Result in an Expanded Phenotype of Okihiro Syndrome Plus Developmental Delay.
Borozdin W, et al.
Human Mutation, 830 null
SALL4 Is a Novel Diagnostic Marker for Testicular Germ Cell Tumors
Cao D, et al.
American Journal of Surgical Pathology, 1065-77 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service