Key Documents
T2567
Dounce tissue grinder tube
working volume 100 mL
Synonym(s):
885303-0100, Dounce Homogenizer, Tissue Grinder, Tissue Homogenizer
About This Item
UNSPSC Code:
41121800
NACRES:
NB.22
material
glass
packaging
pack of 1 ea
working volume
100 mL
General description
Designed primarily for cellular work where the nucleus remains intact after homogenization.
Features and Benefits
• Manufactured from borosilicate glass 3.3• Designed primarily for cellular work where the nucleus remains intact after homogenization.• All-glass construction• Two pestles are supplied with each complete unit• Large clearance pestle is used for the initial sample reduction• Small clearance pestle is used to form the final homogenate• Replacement components are available and completely interchangeable