Skip to Content
Merck
All Photos(2)

Key Documents

SRP0349

Sigma-Aldrich

Flt3 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥80% (SDS-PAGE)

Synonym(s):

CD135, FLK2, STK1, fms-related tyrosine kinase 3

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

54.2 kDa

packaging

pkg of 10 μg

technique(s)

activity assay: suitable
inhibition assay: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... FLT3(2322)

General description

Human Flt3 (GenBank Accession No. NM_004119), amino acids 564-end with N-terminal His-tag, MW=54.2 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Lance et al.
Leukemia, 34(2), 369-379 (2019-08-30)
Three annotated CSF3R mRNA splice variants have been described. CSF3R-V1 is the wild-type receptor, while CSF3R-V4 is a truncated form increased in some patients with AML. CSF3R-V3 mRNA was identified in placenta more than 20 years ago, but remains largely
Connor P Hall et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 22(3), 621-632 (2015-06-18)
The aim of our study is to evaluate the preclinical therapeutic activity and mechanism of action of BEZ235, a dual PI3K/mTOR inhibitor, in combination with dexamethasone in acute lymphoblastic leukemia (ALL). The cytotoxic effects of BEZ235 and dexamethasone as single
Mark Levis et al.
Blood, 106(2), 673-680 (2005-03-31)
Internal tandem duplication mutations of the FLT3 gene (FLT3/ITD mutations) are the most frequent molecular abnormality in acute myeloid leukemia (AML) and are associated with a poor overall survival. While the normal FLT3 receptor is expressed in early hematopoietic progenitor
Shinichiro Takahashi et al.
Leukemia research, 29(8), 893-899 (2005-06-28)
Activating mutations or over-expression of the Flt3 is prevalent in acute myeloblastic leukemia (AML), associated with activation of Ras/MAP kinase and other signaling pathways. In this study, we addressed the role of Flt3 in the activation of nuclear factor-kappa B
Hideki Nakano et al.
Journal of immunology (Baltimore, Md. : 1950), 194(8), 3808-3819 (2015-03-15)
Differential display of the integrins CD103 and CD11b are widely used to distinguish two major dendritic cell (DC) subsets in nonlymphoid tissues. CD103(+) DCs arise from FLT3-dependent DC precursors (preDCs), whereas CD11b(hi) DCs can arise either from preDCs or FLT3-independent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service