Skip to Content
Merck
All Photos(2)

Key Documents

SML3856

Sigma-Aldrich

Alisporivir

≥95% (HPLC)

Synonym(s):

Cyclo[L-alanyl-D-alanyl-N-methyl-L-leucyl-N-methyl-L-leucyl-N-methyl-L-valyl-(2S,3R,4R,6E)-3-hydroxy-4-methyl-2-(methylamino)-6-octenoyl-(2S)-2-aminobutanoyl-N-methyl-D-alanyl-N-ethyl-L-valyl-L-valyl-N-methyl-L-leucyl], DEB025, Debio 025

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C63H113N11O12
CAS Number:
Molecular Weight:
1216.64
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

color

white to off-white

storage temp.

-10 to -25°C

SMILES string

N1([C@H](C(=O)N[C@H](C(=O)N([C@@H](C(=O)N([C@H](C(=O)N[C@H](C(=O)N([C@H](C(=O)N[C@H](C(=O)N[C@@H](C(=O)N([C@H](C(=O)N([C@H](C(=O)N([C@H](C1=O)C(C)C)C)CC(C)C)C)CC(C)C)C)C)C)CC(C)C)C)C(C)C)C(C)C)CC)C)C)CC)[C@H](O)[C@@H](C\C=C\C)C)C

InChI key

OLROWHGDTNFZBH-XEMWPYQTSA-N

Biochem/physiol Actions

Alisporivir is a Cyclophilin A inhibitor with potent anti-hepatitis C virus (HCV) activity. Alisporivir has also been investigated for Duchenne muscular dystrophy.
Cyclophilin A inhibitor; anti-viral with potent anti-hepatitis C virus (HCV) activity

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alisporivir rescues defective mitochondrial respiration in Duchenne muscular dystrophy
Pharmacological Research, 125(Pt B), 122-131 (2017)
DEB025 (Alisporivir) inhibits hepatitis C virus replication by preventing a cyclophilin A induced cis-trans isomerisation in domain II of NS5A
PLoS ONE, 5(10) (2010)
Combating Intracellular Pathogens with Repurposed Host-Targeted Drugs
ACS infectious diseases, 4(2), 88-92 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service