Skip to Content
Merck
All Photos(1)

Key Documents

SML3670

Sigma-Aldrich

Raltegravir

≥98% (HPLC)

Synonym(s):

MK 0518, MK-0518, MK0518, N-[(4-Fluorophenyl)methyl]-1,6-dihydro-5-hydroxy-1-methyl-2-[1-methyl-1-[[(5-methyl-1,3,4-oxadiazol-2-yl)carbonyl]amino]ethyl]-6-oxo-4-pyrimidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21FN6O5
CAS Number:
Molecular Weight:
444.42
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.25

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)CNC(=O)C2N=C(N(C(=O)C2=O)C)C(NC(=O)c3nnc([o]3)C)(C)C

InChI

1S/C20H21FN6O5/c1-10-25-26-17(32-10)16(30)24-20(2,3)19-23-13(14(28)18(31)27(19)4)15(29)22-9-11-5-7-12(21)8-6-11/h5-8,13H,9H2,1-4H3,(H,22,29)(H,24,30)

InChI key

DDGPCENTWMZXFO-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active, potent and selective HIV type-1 (HIV-1) integrase inhibitor in cultures and in vivo.
Raltegravir (MK-0518) is an orally active, potent and selective HIV type-1 (HIV-1) integrase inhibitor (IC50 = 2-7 nM; ineffctive against HIV-1 reverse transcriptase or protease) that prevents HIV-1 cellular replication by blocking viral cDNA integration into host genome in cultures (IC50 = 19 nM & 33 nM in the presence of 10% fetal bovine serum or 50% human serum) and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jamie D Croxtall et al.
Drugs, 68(1), 131-138 (2007-12-18)
* Raltegravir, the first in a new class of orally administered HIV type-1 (HIV-1) integrase inhibitors, selectively inhibits the strand transfer activity of HIV-1 and its integration into human DNA, a key stage in retroviral propagation, thereby limiting viral replication
Antiretroviral activity, pharmacokinetics, and tolerability of MK-0518, a novel inhibitor of HIV-1 integrase, dosed as monotherapy for 10 days in treatment-naive HIV-1-infected individuals
Journal of Acquired Immune Deficiency Syndromes, 43(5), 509-515 (2006)
Pere Domingo et al.
Life sciences, 308, 120948-120948 (2022-09-13)
To assess the potential direct effects of the integrase strand-transfer inhibitors (INsTIs) dolutegravir, bictegravir, and raltegravir, drugs used as treatment for people living with human immunodeficiency virus (PLWH), on human adipose cells. Drugs were added to the differentiation medium of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service