Skip to Content
Merck
All Photos(2)

Key Documents

SML3599

Sigma-Aldrich

IMP-1575

≥98% (HPLC)

Synonym(s):

(+)-(R)-IMP-1575, (+)-IMP-1575, (R)-(+)-2-(Isobutylamino)-1-(4-(6-methylpyridin-2-yl)-6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)ethan-1-one, (R)-1-[6,7-Dihydro-4-(6-methyl-2-pyridinyl)thieno[3,2-c]pyridin-5(4H)-yl]-2-[(2-methylpropyl)amino]ethenone, (R)-IMP-1575

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H25N3OS
Molecular Weight:
343.49
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

oil

color

, Faint yellow to yellow orange

storage temp.

2-8°C

SMILES string

CC1=CC=CC([C@@H]2N(CCC3=C2C=CS3)C(CNCC(C)C)=O)=N1

Biochem/physiol Actions

IMP-1575 is a donor substrate Pal-CoA-competitive (Ki = 38 nM) hedgehog acyltransferase (HHAT) inhibitor with significantly higher potency than its structure analog RUSKI-201 (IC50 = 0.75 vs. 2.0 μM, respectively, by Acyl-cLIP assay).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical Probe Identification of a Small-Molecule Inhibitor Binding Site in Hedgehog Acyltransferase (HHAT)*.
Lanyon-Hogg T, et al.
Angewandte Chemie (International Edition in English), 60, 13542-13547 (2021)
Evaluating Hedgehog Acyltransferase Activity and Inhibition Using the Acylation-coupled Lipophilic Induction of Polarization (Acyl-cLIP) Assay.
Andrei SA, et al.
Methods in Molecular Biology, 2374, 13-26 (2022)
Claire E Coupland et al.
Molecular cell, 81(24), 5025-5038 (2021-12-11)
The Sonic Hedgehog (SHH) morphogen pathway is fundamental for embryonic development and stem cell maintenance and is implicated in various cancers. A key step in signaling is transfer of a palmitate group to the SHH N terminus, catalyzed by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service