Skip to Content
Merck
All Photos(1)

Key Documents

SML3197

Sigma-Aldrich

CA77.1

≥98% (HPLC)

Synonym(s):

N-(4-(6-chloroquinoxalin-2-yl)phenyl)acetamide, N-[4-(6-Chloro-2-quinoxalinyl)phenyl]-acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12ClN3O
CAS Number:
Molecular Weight:
297.74
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Clc1cc2ncc(nc2cc1)c3ccc(cc3)NC(=O)C

InChI

1S/C16H12ClN3O/c1-10(21)19-13-5-2-11(3-6-13)16-9-18-15-8-12(17)4-7-14(15)20-16/h2-9H,1H3,(H,19,21)

InChI key

ZQXMPDVGBWOTBY-UHFFFAOYSA-N

Biochem/physiol Actions

CA77.1 is an orally available, brain penetrant, in vivo active and potent enhancer of chaperone-mediated autophagy (CMA). It improves pathology in Alzheimer′s Disease mouse models. CA77.1 activates CMA in vitro without affecting macroautophagy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service