Skip to Content
Merck
All Photos(1)

Key Documents

SML2770

Sigma-Aldrich

GSK2795039

≥98% (HPLC)

Synonym(s):

GSK 2795039, GSK-2795039, N-(1-Isopropyl-3-(1-methylindolin-6-yl)-1H-pyrrolo[2,3-b]pyridin-4-yl)-1-methyl-1H-pyrazole-3-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N6O2S
CAS Number:
Molecular Weight:
450.56
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=S(NC1=C2C(N(C(C)C)C=C2C3=CC4=C(C=C3)CCN4C)=NC=C1)(C5=NN(C)C=C5)=O

InChI key

FMWVTCZKCXPKFW-UHFFFAOYSA-N

Biochem/physiol Actions

GSK2795039 is a non-cytotoxic (100 μM, 24 hrs, HEK293), NADPH-competitive, potent and selective NOX2 inhibitor (IC50/[NADPH] = 447 nM/25 μM, 2.29 μM/1 mM by NADPH depletion measurement; eNOS & NOX1/3/4/5 IC50 >100 μM, PKCB II IC50 >25 μM, xanthine oxidase IC50 = 28.8 μM). GSK2795039 inhibits PMA-induced reactive oxygen species (ROS) production in cultures (IC50 = 182 nM; HL60) and exhibits in vivo efficacy against complete Freund′s adjuvant-induced paw inflammation (ED50 = 2 mg/kg, ED100 = 100 mg/kg ip.) as well as cerulein-induced acute pancreatitis (ED50 = 100 mg/kg ip.) in mice.
NADPH-competitive, potent and selective NOX2 inhibitor with in vitro and in vivo efficacy against NOX2-mediated ROS production and inflammation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Swenja Kröller-Schön et al.
European heart journal, 39(38), 3528-3539 (2018-06-16)
Aircraft noise causes endothelial dysfunction, oxidative stress, and inflammation. Transportation noise increases the incidence of coronary artery disease, hypertension, and stroke. The underlying mechanisms are not well understood. Herein, we investigated effects of phagocyte-type NADPH oxidase (Nox2) knockout and different
Mavis A A Tenkorang et al.
Endocrinology, 160(4), 947-963 (2019-02-28)
Oxidative stress (OS) is a common characteristic of several neurodegenerative disorders, including Parkinson disease (PD). PD is more prevalent in men than in women, indicating the possible involvement of androgens. Androgens can have either neuroprotective or neurodamaging effects, depending on
Young J Yauger et al.
Journal of neuroinflammation, 16(1), 41-41 (2019-02-20)
Excessive iron contributes to oxidative stress after central nervous system injury. NADPH oxidase (NOX) enzymes are upregulated in microglia after pro-inflammatory activation and contribute to oxidative stress. The relationship between iron, microglia, NOX, and oxidative stress is currently unclear. We
Shannon C Kelly et al.
Redox biology, 27, 101075-101075 (2018-12-24)
The reduction of extracellular oxidants by intracellular electrons is known as trans-plasma membrane electron transport (tPMET). The goal of this study was to characterize a role of tPMET in the sensing of glucose as a physiological signal. tPMET from C2C12
Kazufumi Hirano et al.
Antioxidants & redox signaling, 23(5), 358-374 (2015-07-03)
The NADPH oxidase (NOX) family of enzymes catalyzes the formation of reactive oxygen species (ROS). NOX enzymes not only have a key role in a variety of physiological processes but also contribute to oxidative stress in certain disease states. To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service