Skip to Content
Merck
All Photos(1)

Key Documents

SML1712

Sigma-Aldrich

SBC-115076

≥98% (HPLC)

Synonym(s):

1,5-Dihydro-3-hydroxy-4-[3-methyl-4-(phenylmethoxy)benzoyl]-1-[3-(4-morpholinyl)propyl]-5-(4-pyridinyl)-2H-pyrrol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H33N3O5
CAS Number:
Molecular Weight:
527.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CC1=CC(C(C(C(C2=CC=NC=C2)N3CCCN4CCOCC4)=C(O)C3=O)=O)=CC=C1OCC5=CC=CC=C5

Biochem/physiol Actions

SBC-115076 is an inhibitor of PCSK9 (proprotein convertase subtilisin-kexin 9).
SBC-115076 is an inhibitor of PCSK9 (proprotein convertase subtilisin-kexin 9). PCSK9 binds to the LDL receptor, preventing it from removing LDL cholesterol from the blood. SBC-115076 was shown to inihbit PCSK9, blocking LDLR degradation in HEK293T cells expressing PCSK9 and to increase the uptake of LDL by liver cells in vitro. SBC-115076 lowered cholesterol levels in mice fed a high fat diet.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service