Skip to Content
Merck
All Photos(1)

Key Documents

SML1619

Sigma-Aldrich

Tazarotenic acid

≥98% (HPLC), powder, retinoid receptor (RAR) agonist

Synonym(s):

6-(2-(4,4-Dimethylthiochroman-6-yl)ethynyl)nicotinic acid, 6-[2-(3,4-Dihydro-4,4-dimethyl-2H-1-benzothiopyran-6-yl)ethynyl]-3-pyridinecarboxylic acid, AGN 190299, AGN-190299, Tazarotene acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17NO2S
CAS Number:
Molecular Weight:
323.41
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Tazarotenic acid, ≥98% (HPLC)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1(C)CCSC2=C1C=C(C#CC3=CC=C(C(O)=O)C=N3)C=C2

InChI

1S/C19H17NO2S/c1-19(2)9-10-23-17-8-4-13(11-16(17)19)3-6-15-7-5-14(12-20-15)18(21)22/h4-5,7-8,11-12H,9-10H2,1-2H3,(H,21,22)

InChI key

IQIBKLWBVJPOQO-UHFFFAOYSA-N

Biochem/physiol Actions

Tazarotene is transformed to tazarotenic acid with the help of esterases. Tazarotenic acid regulates the pathogenic aspects of psoriasis through normalizing abnormal keratinocyte differentiation. It possesses strong anti-hyperproliferative effects in skin and reduces inflammation.[1]
Tazarotenic acid, an active metabolite of tazarotene, is a potent and selective agonist of the retinoid receptor (RAR) that binds to RARα, RARβ, and RARγ. Tazarotenic acid relatively selective activates RARβ and RARγ. Tazarotenic acid is a first xenobiotic substrate of human retinoic acid hydroxylase CYP26A1 and CYP26B1.
Tazarotenic acid, an active metabolite of tazarotene, is a potent and selective agonist of the retinoid receptor (RAR).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytochrome P450 2C8 and flavin-containing monooxygenases are involved in the metabolism of tazarotenic acid in humans
Attar M, et al.
Drug Metabolism and Disposition, 31(4), 476-481 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service