Skip to Content
Merck
All Photos(1)

Key Documents

SML1534

Sigma-Aldrich

I-BRD9

≥98% (HPLC)

Synonym(s):

N-(1,1-Dioxidotetrahydro-2H-thiopyran-4-yl)-5-ethyl-4-oxo-7-(3-(trifluoromethyl)phenyl)-4,5-dihydrothieno[3,2-c]pyridine-2-carboximidamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H22F3N3O3S2
CAS Number:
Molecular Weight:
497.55
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

CCN1C(C2=C(SC(C(NC3CCS(CC3)(=O)=O)=N)=C2)C(C4=CC(C(F)(F)F)=CC=C4)=C1)=O

InChI

1S/C22H22F3N3O3S2/c1-2-28-12-17(13-4-3-5-14(10-13)22(23,24)25)19-16(21(28)29)11-18(32-19)20(26)27-15-6-8-33(30,31)9-7-15/h3-5,10-12,15H,2,6-9H2,1H3,(H2,26,27)

InChI key

WRUWGLUCNBMGPS-UHFFFAOYSA-N

General description

I-BRD9 is developed from thienopyridone scaffold. It can also be used as an identifier for BRD9 regulated gene in Kasumi-1 cells involved in oncology and immune response pathways.

Biochem/physiol Actions

I-BRD9 is a selective cellular chemical probe for bromodomain-containing protein 9 (BRD9), thought to be a member of the chromatin remodelling SWI/SNF BAF complex, which plays a fundamental role in gene expression regulation. I-BRD9 has a pIC50 value of 7.3 with greater than 700-fold selectivity over the BET family and 200-fold over the highly homologous bromodomain-containing protein 7 (BRD7) and greater than 70-fold selectivity against a panel of 34 bromodomains. For full characterization details, please visit the I-BRD9 probe summary on the Structural Genomics Consortium (SGC) website.

To learn about other SGC chemical probes for epigenetic targets, visit sigma.com/sgc
I-BRD9 is a selective cellular chemical probe for bromodomain-containing protein 9 (BRD9).

Features and Benefits

I-BRD9 is an epigenetic chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.
This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

I-BRD9 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the I-BRD9 probe summary on the Chemical Probes Portal website.

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natalie H Theodoulou et al.
Journal of medicinal chemistry, 59(4), 1425-1439 (2015-04-10)
Acetylation of histone lysine residues is one of the most well-studied post-translational modifications of chromatin, selectively recognized by bromodomain "reader" modules. Inhibitors of the bromodomain and extra terminal domain (BET) family of bromodomains have shown profound anticancer and anti-inflammatory properties
Structure-based design of an in vivo active selective BRD9 inhibitor.
Martin L J, et al.
Journal of Medicinal Chemistry, 59(10), 4462-4475 (2016)

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service