Skip to Content
Merck
All Photos(1)

Key Documents

SML1491

Sigma-Aldrich

GW501516

≥98% (HPLC)

Synonym(s):

2-[2-Methyl-4-[[[4-methyl-2-[4-(trifluoromethyl)phenyl]-5-thiazolyl]methyl]thio]phenoxy]-acetic acid, GW1516

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18F3NO3S2
CAS Number:
Molecular Weight:
453.50
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(F)(F)c1ccc(cc1)c2[s]c(c(n2)C)CSc3cc(c(cc3)OCC(=O)O)C

InChI

1S/C21H18F3NO3S2/c1-12-9-16(7-8-17(12)28-10-19(26)27)29-11-18-13(2)25-20(30-18)14-3-5-15(6-4-14)21(22,23)24/h3-9H,10-11H2,1-2H3,(H,26,27)

InChI key

YDBLKRPLXZNVNB-UHFFFAOYSA-N

Application

GW501516 has been used as a peroxisome proliferator-activated receptor (PPAR) δ agonist:
  • to treat the differentiated L6 myotubes to test its effect on free fatty acid (FFA) uptake under mild hypoxia conditions
  • in organoid culture medium to test its effect on self-renewal of PR domain containing 16 (Prdm16)-depleted intestinal stem cells
  • as a component in the defatting medium for primary human hepatocytes (PHH) culture

GW501516 has been used to study its effect on mutation-driven colorectal tumorigenesis and tumor invasion using mouse model.

Biochem/physiol Actions

GW501516 is a selective Peroxisome Proliferator-Activated Receptor delta (PPARδ) agonist with high affinity (Ki = 1 nM) and potency (EC50 = 1 nM) for PPARδ and > 1000 fold selectivity over PPARα and PPARγ. GW501516 was shown to activate some of the same pathways activated through exercise, and was investigated as a potential treatment for obesity, diabetes, dyslipidemia and cardiovascular disease. However, GW501516 also showed an increase in cancer in rats.
GW501516 is a selective Peroxisome Proliferator-Activated Receptor delta (PPARδ) agonist.
PPARδ activation by GW501516, retards weight gain through fatty acid catabolism in adipose tissue and skeletal muscles. GW501516 causes an increase in the levels high-density lipoprotein cholesterol and apolipoprotein A (apoA) and reduction in the low density-lipoprotein cholesterol, apoB, and triglyceride.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid effects of peroxisome proliferator-activated receptor-δ agonist GW501516 in subjects with low high-density lipoprotein cholesterol: characteristics of metabolic syndrome.
Olson E J, et al.
Arteriosclerosis, Thrombosis, and Vascular Biology, 32(9), 2289-2294 (2012)
Activation of peroxisome proliferator-activated receptor-δ by GW501516 prevents fatty acid-induced nuclear factor-κB activation and insulin resistance in skeletal muscle cells.
Coll T, et al.
Endocrinology, 151(4), 1560-1569 (2010)
Pleotropic effects of PPARD accelerate colorectal tumor progression and invasion.
Liu Y, et al.
bioRxiv, 325001-325001 (2018)
Yi Ling Low et al.
Molecular pharmaceutics, 17(3), 873-884 (2020-01-17)
Brain levels of docosahexaenoic acid (DHA), an essential cognitively beneficial fatty acid, are reduced in Alzheimer's disease (AD). We have demonstrated in an AD mouse model that this is associated with reduced blood-brain barrier (BBB) transport of DHA and lower
Buqing Ye et al.
The EMBO journal, 39(13), e103786-e103786 (2020-05-26)
Lgr5+ intestinal stem cells (ISCs) exhibit self-renewal and differentiation features under homeostatic conditions, but the mechanisms controlling Lgr5 + ISC self-renewal remain elusive. Here, we show that the chromatin remodeler SRCAP is highly expressed in mouse intestinal epithelium and ISCs. Srcap deletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service