Skip to Content
Merck
All Photos(1)

Key Documents

SML1403

Sigma-Aldrich

IN-1130

≥98% (HPLC)

Synonym(s):

3-[[5-(6-Methyl-2-pyridinyl)-4-(6-quinoxalinyl)-1H-imidazol-2-yl]methyl]-benzamide, IN 1130

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H20N6O
CAS Number:
Molecular Weight:
420.47
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[nH]1c(nc(c1c4cc5nccnc5cc4)c3nc(ccc3)C)Cc2cc(ccc2)C(=O)N

InChI

1S/C25H20N6O/c1-15-4-2-7-20(29-15)24-23(17-8-9-19-21(14-17)28-11-10-27-19)30-22(31-24)13-16-5-3-6-18(12-16)25(26)32/h2-12,14H,13H2,1H3,(H2,26,32)(H,30,31)

InChI key

RYKSGWSKILPDDY-UHFFFAOYSA-N

Biochem/physiol Actions

IN-1130 is a potent inhibitor of the activin receptor-like kinase ALK-5 (Type 1 TGF1β recptor). The compound inhibits phosphorylation of Smad-2 with an IC50 of 36 nM. IN-1130 inhibits renal fibrosis, and also reduces tumor volume in prostate tumor xenografts in murine models.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service