Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502580

Sigma-Aldrich

Anti-Retinoic Acid Receptor α antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NR1B1, Nuclear receptor subfamily 1 group B member 1, RAR-α, RARA, Retinoic acid receptor α

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RARA(5914)

General description

Anti-Retinoic Acid Receptor α Antibody detects endogenous levels of total Retinoic Acid Receptor α protein.
Retinoic acid receptor α (RARα) is a nuclear receptor for retinoic acid and is expressed in the retina. It belongs to the nuclear steroid/thyroid hormone receptor superfamily. The gene encoding it is localized on human chromosome 17q21.

Immunogen

The antiserum was produced against synthesized peptide derived from human Retinoic Acid Receptor alpha.

Immunogen Range: 46-95

Biochem/physiol Actions

Retinoic acid receptor α (RARα) is a ligand-dependent transcriptional modulator of genes that has a role in differentiation, cellular development and apoptosis. It has a role in germ-cell development in testis. The protein may be associated with acute promyelocytic leukemia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activity of retinoic acid receptor-alpha is directly regulated at its protein kinase A sites in response to follicle-stimulating hormone signaling.
Santos NC and Kim KH
Endocrinology, 151(5), 2361-2372 (2010)
Enhanced expression of retinoic acid receptor alpha (RARA) induces epithelial-to-mesenchymal transition and disruption of mammary acinar structures.
Doi A
Molecular Oncology, 9(2), 355-364 (2015)
The retinoic acid receptor alpha (RARA) gene is not associated with myopia, hypermetropia, and ocular biometric measures.
Veerappan S
Molecular Vision, 15, 1390-1397 (2009)
NLS-RARa promotes proliferation and inhibits differentiation in HL-60 cells.
Hu XX
International Journal of Medical Sciences, 11(3), 247-254 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service