Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500835

Sigma-Aldrich

Anti-PYK2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CADTK, CAK β, Calcium-dependent tyrosine kinase, Cell adhesion kinase β, FADK 2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 115 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PTK2B(2185)

General description

Protein tyrosine kinase 2 β (PTK2B) is a member of the focal adhesion kinase family which is expressed in the brain. The gene encoding it is localized on human chromosome 8.

Immunogen

The antiserum was produced against synthesized peptide derived from human PYK2.

Immunogen Range: 546-595

Biochem/physiol Actions

Protein tyrosine kinase 2 β (PTK2B) takes part in the β2-mediated signaling pathway and activates mitogen activated protein kinase pathway. The protein has been shown to be upregulated in many tumors. PTK2B also has a role in cell proliferation and migration. It regulates ion channels which are induced by calcium.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Steblyanko et al.
The Journal of biological chemistry, 290(19), 11833-11842 (2015-03-18)
Integrin engagement on lymphocytes initiates "outside-in" signaling that is required for cytoskeleton remodeling and the formation of the synaptic interface. However, the mechanism by which the "outside-in" signal contributes to receptor-mediated intracellular signaling that regulates the kinetics of granule delivery
Heval M Ozgen et al.
Journal of autism and developmental disorders, 39(2), 322-329 (2008-08-13)
Autism spectrum disorders (ASDs) are a group of neurodevelopmental disorders with a strong genetic etiology. Cytogenetic abnormalities have been detected in 5-10% of the patients with autism. In this study, we present the clinical, cytogenetic and array-comparative genomic hybridization (array-CGH)
Elinor Y Gottschalk et al.
Journal of virology, 89(17), 9103-9114 (2015-06-26)
The infectious process of human papillomaviruses (HPVs) has been studied considerably, and many cellular components required for viral entry and trafficking continue to be revealed. In this study, we investigated the role of the nonreceptor tyrosine kinase Pyk2 during HPV16
Yu Zhang et al.
Blood, 124(17), 2675-2686 (2014-09-14)
Proline-rich tyrosine kinase 2 (Pyk2) is a member of the focal adhesion kinase family that has been recently linked to tumor development. However, its role in modulating multiple myeloma (MM) biology and disease progression remains unexplored. We first demonstrated that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service