Skip to Content
Merck
All Photos(1)

Key Documents

SAB2501424

Sigma-Aldrich

Anti-IGF1 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-IGF1A, Anti-IGFI, Anti-OTTHUMP00000195084, Insulin-like growth factor 1, Insulin-like growth factor 1 (somatomedin C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGF1(3479)

General description

Insulin-like growth factor 1 (IGF1) is a single polypeptide chain protein with three disulfide bridges. It is mainly produced by the liver and a wide range of extra-hepatic tissues. IGF1 is seen in normal and neoplastic breasts. The IGF1 gene is 84779 bp long and is made of 6 different exons. IGF-1 gene is located on human chromosome 12q23.2.

Immunogen

Peptide with sequence RSVRAQRHTD, from the internal region of the protein sequence according to NP_001104753.1; NP_001104754.1; NP_001104755.1; NP_000609.1

Application

Anti-IGF1 antibody produced in goat has been used in western blotting.

Biochem/physiol Actions

Insulin-like growth factor 1 (IGF1) protein is essential for the development of children and adults. It plays a crucial role in general anabolic processes. IGF1 participates in differentiation and cell proliferation. It plays a key role in osteogenesis, muscle repair, and hypertrophy after trauma. IGF1 is also involved in the generation of axons in nerves and regeneration of a nerve after ischemic insult. It is known to participate in the clinical sequelae of Laron′s dwarfism. IGF1 is required for normal breast development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IGF1 (Insulin-Like Growth Factor 1 (Somatomedin C))
Panayi ND and Burd R
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2013)
M Ouni et al.
The pharmacogenomics journal, 16(1), 102-106 (2015-04-15)
Short children using growth hormone (GH) to accelerate their growth respond to this treatment with a variable efficacy. The causes of this individual variability are multifactorial and could involve epigenetics. Quantifying the impact of epigenetic variation on response to treatments
T L Wise
Genes, brain, and behavior, 16(2), 241-249 (2016-09-20)
Fragile X syndrome (FXS) is an inherited form of intellectual disability that is usually caused by expansion of a polymorphic CGG repeat in the 5' untranslated region of the X-linked FMR1 gene, which leads to hypermethylation and transcriptional silencing. Two
Marina Valente Barroso et al.
Bioorganic & medicinal chemistry, 25(20), 5557-5568 (2017-09-10)
Chronic obstructive pulmonary disease (COPD) is an incurable and progressive disease. Emphysema is the principal manifestation of COPD, and the main cause of this condition is cigarette smoke (CS). Natural products have shown antioxidant and anti-inflammatory properties that can prevent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service