Skip to Content
Merck
All Photos(1)

Key Documents

SAB1406820

Sigma-Aldrich

Anti-TMPRSS11D antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

HAT, MGC150587, MGC150588

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~46.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

This gene encodes a trypsin-like serine protease released from the submucosal serous glands onto mucous membrane. It is a type II integral membrane protein and has 29-38% identity in the sequence of the catalytic region with human hepsin, enteropeptidase, acrosin, and mast cell tryptase. The noncatalytic region has little similarity to other known proteins. This protein may play some biological role in the host defense system on the mucous membrane independently of or in cooperation with other substances in airway mucous or bronchial secretions. (provided by RefSeq)

Immunogen

TMPRSS11D (NP_004253.1, 1 a.a. ~ 418 a.a) full-length human protein.

Sequence
MYRPARVTSTSRFLNPYVVCFIVVAGVVILAVTIALLVYFLAFDQKSYFYRSSFQLLNVEYNSQLNSPATQEYRTLSGRIESLITKTFKESNLRNQFIRAHVAKLRQDGSGVRADVVMKFQFTRNNNGASMKSRIESVLRQMLNNSGNLEINPSTEITSLTDQAAANWLINECGAGPDLITLSEQRILGGTEAEEGSWPWQVSLRLNNAHHCGGSLINNMWILTAAHCFRSNSNPRDWIATSGISTTFPKLRMRVRNILIHNNYKSATHENDIALVRLENSVTFTKDIHSVCLPAATQNIPPGSTAYVTGWGAQEYAGHTVPELRQGQVRIISNDVCNAPHSYNGAILSGMLCAGVPQGGVDACQGDSGGPLVQEDSRRLWFIVGIVSWGDQCGLPDKPGVYTRVTAYLDWIRQQTGI

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service