RAB0394
HumanPDGF-AA ELISA Kit
for serum, plasma, cell culture supernatant and urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type plasma
sample type serum
sample type urine
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 40 pg/mL
standard curve range: 41.15-30000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... PDGFA(5154)
General description
The Human PDGF-AA (human platelet-derived growth factor AA) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human PDGF-AA in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human PDGF-AA
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
- RABDIL6ELISA 5X Sample Diluent Buffer (Item D2)SDS
- RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS
- RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS
- RABSTOP3ELISA Stop Solution (Item I)SDS
- RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS
- RABWASH420X Wash Buffer (Item B)SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Apheresis Platelet Rich-Plasma for Regenerative Medicine: An In Vitro Study on Osteogenic Potential.
International journal of molecular sciences, 22(16) (2021-08-28)
Background: Platelet-Rich Plasma (PRP) induces bone regeneration; however, there is low evidence supporting its efficacy in bone healing. The lack of a standardized protocol of administration represents the main obstacle to its use in the clinical routine for bone defects'
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service