Skip to Content
Merck
All Photos(2)

Key Documents

RAB0008

Sigma-Aldrich

Mouse RAGE / AGER ELISA Kit

for serum, plasma and cell culture supernatant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
capture ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 13.72-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Ager(11596)

General description

The Mouse RAGE (Receptor for Advanced Glycation End Products) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse RAGE in serum, plasma, cell culture supernatants and urine.

Immunogen

Recombinant Mouse RAGE

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABELADEELISA 5X Assay/Sample Diluent Buffer E (Item E2)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David G S Farmer et al.
Pulmonary pharmacology & therapeutics, 29(1), 7-14 (2014-01-15)
The calgranulin-like protein MTS1/S100A4 and the receptor for advanced glycation end-products (RAGE) have recently been implicated in mediating pulmonary arterial smooth muscle cell proliferation and vascular remodelling in experimental pulmonary arterial hypertension (PH). Here, the effects of RAGE antagonism upon
Yin-Ching Chuang et al.
BMC pulmonary medicine, 14, 115-115 (2014-07-16)
Glutamine (GLN) has been reported to improve clinical and experimental sepsis outcomes. However, the mechanisms underlying the actions of GLN remain unclear, and may depend upon the route of GLN administration and the model of acute lung injury (ALI) used.
Yuuki Shimizu et al.
Journal of the American Heart Association, 9(4), e014691-e014691 (2020-02-19)
Background DJ-1 is a ubiquitously expressed protein typically associated with the development of early onset Parkinson disease. Recent data suggest that it also plays a role in the cellular response to stress. Here, we sought to determine the role DJ-1
Zhi-hong Wang et al.
Molecular nutrition & food research, 58(7), 1426-1435 (2014-03-04)
Antiglycative effects of glycyrrhizic acid (GA) in kidney of diabetic mice were examined. GA at 0.05, 0.1, and 0.2% was supplied to diabetic mice for 9 wk. Results showed that GA intake increased its deposit in kidney, raised plasma insulin
Anisuzzaman et al.
The Journal of clinical investigation, 124(10), 4429-4444 (2014-11-18)
Ticks are notorious hematophagous ectoparasites and vectors of many deadly pathogens. As an effective vector, ticks must break the strong barrier provided by the skin of their host during feeding, and their saliva contains a complex mixture of bioactive molecules

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service