Skip to Content
Merck
All Photos(1)

Key Documents

N6784

Sigma-Aldrich

Noggin/Fc Chimera from mouse

>95% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.32

biological source

mouse

Quality Level

recombinant

expressed in NSO cells

Assay

>95% (SDS-PAGE)

form

lyophilized powder

mol wt

55-60 kDa by SDS-PAGE (reducing)
calculated mol wt 50 kDa

packaging

pkg of 50 μg

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Nog(18121)

Biochem/physiol Actions

Binds and blocks the acitivity of bone morphogenetic proteins (BMPs). Mature mouse noggin shares 99% and 83% sequence identity with human and Xenopus noggin, respectively.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline.

Analysis Note

The biological activity is measured by its ability to inhibit recombinant human BMP-4 induced alkaline phosphatase activity in ATDC5 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Noggin antagonism of BMP4 signaling controls development of the axial skeleton in the mouse.
Wijgerde M
Developmental Biology (2005)
TGF-beta superfamily members and ovarian follicle development.
Knight PG and Glister C
Reproduction (Cambridge, England) (2006)
A H Reddi
Arthritis research, 3(1), 1-5 (2001-02-15)
This commentary is a concise discussion of the interactions between bone morphogenetic proteins (BMPs) and their binding proteins in bone and cartilage morphogenesis. BMPs are a family of growth and differentiation factors, and they act on mesenchymal cells to induce
Human disease-causing NOG missense mutations: effects on noggin secretion, dimer formation, and bone morphogenetic protein binding.
Marcelino J
Proceedings of the National Academy of Sciences of the USA (2001)
Impaired osteoblastic differentiation, reduced bone formation, and severe osteoporosis in noggin-overexpressing mice.
Wu XB
The Journal of Clinical Investigation (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service