MTCP3801QW
MTC™ Bio Pure.Amp™qPCR 8 Tube Strips
capacity 0.1 mL, white tube, separate, optical clear flat cap
Synonym(s):
PCR sample storage tubes, plastic tube, sample storage tube
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
Recommended Products
material
clear cap (optical)
clear tube
flat cap
polypropylene
white tube
sterility
non-sterile
packaging
pack of 120 ea
manufacturer/tradename
MTC™ Bio P3801-QW
capacity
0.1 mL
General description
0.1ml qPCR 8-Strip (With SEPARATE Optical Strip Caps), WHITE 120/pk
Pure.Amp™ 0.1mL qPCR strips strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer and are certified free of RNase, DNase, DNA and PCR inhibitors, and are made of heat-resistant medical-grade polypropylene.
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)