Skip to Content
Merck
All Photos(1)

Key Documents

L5016

Sigma-Aldrich

1-O-Palmityl-sn-glycero-3-phosphocholine

≥99%, synthetic

Synonym(s):

1-Hexadecyl-sn-glycero-3-phosphocholine, 3-sn-Lysophosphatidylcholine, 1-hexadecyl, Lyso-PAF-C16, Lyso-platelet activating factor

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H52NO6P
CAS Number:
Molecular Weight:
481.65
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

Quality Level

Assay

≥99%

storage temp.

−20°C

SMILES string

O[C@](COP([O-])(OCC[N+](C)(C)C)=O)([H])COCCCCCCCCCCCCCCCC

InChI

1S/C24H52NO6P/c1-5-6-7-8-9-10-11-12-13-14-15-16-17-18-20-29-22-24(26)23-31-32(27,28)30-21-19-25(2,3)4/h24,26H,5-23H2,1-4H3/t24-/m1/s1

InChI key

VLBPIWYTPAXCFJ-XMMPIXPASA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Intermediate in the synthesis of platelet activating factor (PAF). Inactive form that can be used as a control in studies of PAF activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Robinson et al.
Biochimica et biophysica acta, 837(1), 52-56 (1985-10-23)
1-Alkyl-2-acetyl-sn-glycero-3-phosphocholine (alkylacetyl-GPC; platelet-activating factor; PAF) is actively taken up and metabolized by rat alveolar macrophages maintained in culture. The major metabolic products are lyso-PAF (alkyllyso-GPC) and alkylacyl-GPC. Lyso-PAF accumulates primarily in the media, whereas alkylacyl-GPC is predominantly associated with cellular
Sturk, A., et al.
Journal of Lipid Research, 23, 219-219 (1989)
C G Qian et al.
Journal of lipid mediators, 1(2), 113-123 (1989-03-01)
Suspensions of neonatal rat myocytes were used to investigate the metabolism of [3H]PAF (1-alkyl-2-acetyl-(sn-glycero-3-phosphocholine) (GPC] and [3H]alkyllyso-GPC. [3H]Alkylacyl-GPC consisting of molecular species with four or more double bonds (87%) was the major metabolite formed when either [3H]PAF (4 x 10(-6)
J K Mickelson et al.
Journal of molecular and cellular cardiology, 20(6), 547-561 (1988-06-01)
Myocardial injury was produced in separated groups of anesthetized rabbits by occlusion of the left circumflex coronary artery for 1 h followed by reperfusion for 2, 4, or 6 h after release of the occlusive ligature. The ischemically-injured and reperfused
Kimberly D Dyer et al.
Journal of immunology (Baltimore, Md. : 1950), 184(11), 6327-6334 (2010-04-28)
Platelet-activating factor (PAF [1-O-alkyl-2-acetyl-sn-glycero-3-phosphocholine]) is a phospholipid mediator released from activated macrophages, mast cells, and basophils that promotes pathophysiologic inflammation. Eosinophil responses to PAF are complex and incompletely elucidated. We show in this article that PAF and its 2-deacetylated metabolite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service