Skip to Content
Merck
All Photos(1)

Key Documents

I7642

Sigma-Aldrich

Anti-Interleukin-12 antibody produced in goat

IgG fraction of antiserum

Synonym(s):

Anti-IL-12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

technique(s)

neutralization: suitable
western blot: 1-2 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Il12a(16159)

General description

Interleukin-12 (IL-12) is an important cytokine that links innate and adaptive immunity. It plays a role in the regulation of Th1-associated cellular immunity to foreign pathogens, and humoral immunity to antigens that are both T cell-independent and T cell-dependent. IL-12 is primarily produced by innate immune cells, dendritic cells, epithelial cells and macrophages. All forms of IL-12 bind to the IL-12R which includes IL-12RbI and IL-12RbII subunits expressed on T cells and natural cells. IL-12 stimulates the production of IFN-γ and TNF-α, subsequent production of nitric oxide that leads to pathogen destruction. Reports suggest the involvement of JAK/STAT pathway components in mediating these effects. The effects of IL-12 have been implicated in bacterial, fungal and viral infections. Due to its role in inducing matrix metalloproteases, it also has important role in the etiology of chronic B-cell malignancies and lung adenocarcinomas
Anti-Interleukin-12 antibody recognizes mouse IL-12. It shows < 10% cross-reactivity with recombinant human IL-12 and < 5% cross-reactivity with recombinant porcine IL-12.

Specificity

The antibody will neutralize the biological activity of recombinant mouse IL-12. Based on ELISA, the antibody shows ~10% cross-reactivity with recombinant human IL-12 and <5% cross-reactivity with recombinant porcine IL-12.

Immunogen

purified, insect cell line Sf21-derived, recombinant mouse IL-12.

Application

Anti-Interleukin-12 antibody may be used for immunoblotting at a working concentration of 1-2 μg/ml. The antibody is also suitable for neutralization reactions. It has been used to neutralize IL-12 in the extracts of murine splenocytes.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline containing carbohydrates.

Preparation Note

Purified using protein G.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ludwig Deml et al.
Infection and immunity, 73(8), 4732-4742 (2005-07-26)
Predominant T-helper 1 (Th1) responses with increased gamma interferon (IFN-gamma) levels have been proposed to play an important role in Helicobacter pylori-induced gastritis and peptic ulceration. However, bacterial factors contributing to the initiation of Th1 polarization of H. pylori-specific immune
IL-12 and Related Cytokines: Function and Regulatory Implications in Candida albicans Infection
Ashman RB et al
Clinical & Developmental Immunology, doi: 10-doi: 10 (2011)
Anne O'Garra et al.
Nature immunology, 10(9), 929-932 (2009-08-21)
The authors recount their discovery of how pathogen-induced interleukin 12 production leads to T(H)1 T cell polarization. Simultaneously they discovered the suppressive cytokine interleukin 10 inhibits antigen-presenting cells, thus regulating development of T(H)1 cells.
Dennis W Metzger
Cytokine, 52(1-2), 102-107 (2010-07-24)
Interleukin (IL)-12 is a pivotal cytokine that strongly stimulates Th1-associated cellular immunity. It is now recognized that IL-12 also activates humoral immunity to both T-dependent and T-independent antigens. This has let to considerable interest in exploiting IL-12 as a vaccine
David J Klinke
Molecular cancer, 9, 242-242 (2010-09-17)
Monoclonal antibodies represent some of the most promising molecular targeted immunotherapies. However, understanding mechanisms by which tumors evade elimination by the immune system of the host presents a significant challenge for developing effective cancer immunotherapies. The interaction of cancer cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service