Key Documents
HS24490D
LabAlert® Pocket Timer/Stopwatch
4 Channel, timing 100 hr, resolution: 1/100 sec
About This Item
UNSPSC Code:
41122411
NACRES:
NB.35
Pricing and availability is not currently available.
description
alarm
timing measuring range
100 hr, resolution: 1/100 sec
L × H × D
61 × 71 × 25 mm
battery power
1 × 1.5 V (LR44)
Related Categories
General description
Pocket, four channel stopwatch / timer with both audible and visual alarms.
Features and Benefits
  • Four independent timing channels with visual and audible alarms
  • use as a stopwatch, a count up/down, or a clock
  • includes spring clip and magnetic back
Legal Information
Clearly Safe is a registered trademark of Heathrow Scientific LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)