Skip to Content
Merck
All Photos(4)

Key Documents

HPA040668

Sigma-Aldrich

Anti-SLC8B1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ22233, Anti-NCKX6, Anti-NCLX, Anti-SLC24A6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

RQRRGSLFCPMPVTPEILSDSEEDRVSSNTNSYDYGDEYRPLFFYQETTAQILVRALNPLDYMKWRRKSAYWKAL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC24A6(80024)

General description

Solute carrier family 8 member B1 (SLC8B1), also known as SLC24A6 or NCLX (mitochondrial Na/Ca exchanger) is encoded by the gene mapped to human chromosome 12q24.13. The encoded protein belongs to the SLC family and is expressed in variety tissues including vessel, lymph node, ovary, bladder and testis.

Immunogen

solute carrier family 24 (sodium/potassium/calcium exchanger), member 6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 8 member B1 (SLC8B1) functions as a mitochondrial sodium calcium exchanger and is implicated in maintaining cellular Ca2+ homeostasis in diverse tissues and cell types. The encoded protein regulates Ca2+ induced NAD(P)H production and matrix redox state by controlling the mitochondrial Ca2+ elevations. SLC8B1 also plays an essential role in regulating automaticity of the HL-1 cardiomyocytes by controlling sarcoplasmic reticulum (SR) Ca2+ handling .

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST80981

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinjiang Cai et al.
The Journal of biological chemistry, 279(7), 5867-5876 (2003-11-20)
Bioinformatic and molecular cloning tools were used to identify and isolate cDNA clones from mouse and human tissues that encode the sixth member of the K(+)-dependent Na+/Ca2+ exchanger family, NCKX6. The mouse NCKX6 protein is 585 amino acids long and
Umberto De Marchi et al.
The Journal of biological chemistry, 289(29), 20377-20385 (2014-06-06)
Mitochondria capture and subsequently release Ca(2+) ions, thereby sensing and shaping cellular Ca(2+) signals. The Ca(2+) uniporter MCU mediates Ca(2+) uptake, whereas NCLX (mitochondrial Na/Ca exchanger) and LETM1 (leucine zipper-EF-hand-containing transmembrane protein 1) were proposed to exchange Ca(2+) against Na(+)
Ayako Takeuchi et al.
Scientific reports, 3, 2766-2766 (2013-09-27)
Mitochondrial Ca(2+) is known to change dynamically, regulating mitochondrial as well as cellular functions such as energy metabolism and apoptosis. The NCLX gene encodes the mitochondrial Na(+)-Ca(2+) exchanger (NCXmit), a Ca(2+) extrusion system in mitochondria. Here we report that the
Maxime Guéguinou et al.
Cellular and molecular life sciences : CMLS, 79(6), 284-284 (2022-05-09)
Recent evidences highlight a role of the mitochondria calcium homeostasis in the development of colorectal cancer (CRC). To overcome treatment resistance, we aimed to evaluate the role of the mitochondrial sodium-calcium-lithium exchanger (NCLX) and its targeting in CRC. We also
Xianren Wang et al.
Frontiers in molecular neuroscience, 11, 469-469 (2019-01-24)
Mitochondria modulate cellular calcium homeostasis by the combined action of the mitochondrial calcium uniporter (MCU), a selective calcium entry channel, and the sodium calcium exchanger (NCLX), which extrudes calcium from mitochondria. In this study, we investigated MCU and NCLX in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service