Skip to Content
Merck
All Photos(9)

Key Documents

HPA025735

Sigma-Aldrich

Anti-ACOT7 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-Acyl-CoA thioesterase 7, Anti-CTE-II, Anti-CTE-IIa, Anti-Cytosolic acyl coenzyme A thioester hydrolase, Anti-Long chain acyl-CoA thioester hydrolase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

RLCEFGRQASSRRLVAGQGCVGPRRGCCAPVQVVGPRADLPPCGACITGRIMRPDDANVAGNVHGGTILKMIEEAGAIISTRHCNSQNGE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACOT7(11332)

General description

The gene ACOT7 (acyl-CoA thioesterase 7) is highly expressed in the brain and testis, and is mapped to human chromosome 1p36. This gene produces many isoforms due to alternate splicing. The encoded protein is present in the cytoplasm and nucleus. It belongs to acyl-CoA hydrolases class of proteins. These proteins are mainly responsible for hydrolysis of acyl-CoA thioesters to free fatty acids and CoA-SH.

Immunogen

Cytosolic acyl coenzyme A thioester hydrolase recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

ACOT7 (acyl-CoA thioesterase 7) is involved in fatty acid oxidation. It is a connecting link between metabolism of acyl-CoAs and cholesterol in neurons. Overexpression of this gene in macrophage cell line modulates the generation of prostaglandins D2 and E2, thereby making it a drug target in inflammatory diseases. ACOT7 has high specificity for arachido-258 noyl-CoA (AA-CoA). It might also be associated with mesial temporal lobe epilepsy (MTLE).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76570

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Human brain acyl-CoA hydrolase isoforms encoded by a single gene.
Yamada J
Biochemical and Biophysical Research Communications, 299, 49-56 (2002)
Structural basis for recruitment of tandem hotdog domains in acyl-CoA thioesterase 7 and its role in inflammation.
Forwood JK
Proceedings of the National Academy of Sciences of the USA, 104, 10382-10387 (2007)
Sterol Regulatory Element-Binding Protein-2 modulates human brain acyl-CoA hydrolase gene transcription.
Takagi M
Molecular and Cellular Biochemistry, 275, 199-206 (2005)
Functional and structural properties of mammalian acyl-coenzyme A thioesterases.
Kirkby B
Progress in Lipid Research, 49, 366-377 (2010)
Aberrant cytosolic acyl-CoA thioester hydrolase in hippocampus of patients with mesial temporal lobe epilepsy.
Yang JW
Amino Acids, 27, 269-275 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service