Skip to Content
Merck
All Photos(3)

Key Documents

HPA015027

Sigma-Aldrich

Anti-ADGRL3 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-KIAA0768, Anti-LEC3, Anti-LPHN3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

DNLLYVWNNYHVVKYSLDFGPLDSRSGQAHHGQVSYISPPIHLDSELERPSVKDISTTGPLGMGSTTTSTTLRTTTLSPGRSTTPSVSGRRNRSTSTPSPAVEVLDDMTTHLPSASSQIPALEESCEAVEAREIMWFKTRQGQIAKQPCP

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LPHN3(23284)

General description

LPHN3 (latrophilin3) is a G-protein couple receptor (GRPC) belonging to the LPHN family. It is the major LPHN member expressed in brain, especially in cerebellum, cerebral cortex, amygdala, and caudate nucleus. This gene is localized to human chromosome 4q. It has a 19-amino acid leader peptide in its long N-terminal exoplasmic region, and it spans the membrane seven times. It also contains a glycosylation region rich in serine/threonine.

Immunogen

adhesion G protein-coupled receptor L3

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The exact functional characteristics of LPHN3 (latrophilin3) are not yet understood. In vitro studies show that this protein acts as a receptor for fibronectin leucine-rich repeat transmembrane (FLRT) protein, and this complex might be involved in the glutamatergic synapse development. SNPs in this gene influence metabolism in neuronal circuits, which are involved in attention-deficit/hyperactivity disorder (ADHD). Variants in this gene are associated with susceptibility to ADHD. Studies show that elevated LPHN3 mRNA expression in breast cancer is associated with axillary-node metastasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72455

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Acosta et al.
Translational psychiatry, 1, e17-e17 (2011-01-01)
The severity of attention-deficit/hyperactivity disorder (ADHD) symptoms is a major predictor of long-term ADHD outcome. To investigate if two-locus interactions might predict ADHD severity, we studied a sample of 1341 individuals from families clustering ADHD, using the Vanderbilt Assessment Scale
Andreas J Fallgatter et al.
European neuropsychopharmacology : the journal of the European College of Neuropsychopharmacology, 23(6), 458-468 (2012-12-19)
Current research strategies have made great efforts to further elucidate the complex genetic architecture of attention-deficit hyperactivity disorder (ADHD). The present study examined the impact of an LPHN3 haplotype that has recently been associated with ADHD (Arcos-Burgos et al., 2010)
Manas Kotepui et al.
Asian Pacific journal of cancer prevention : APJCP, 13(11), 5879-5882 (2013-01-16)
Breast cancer is the leading cause of cancer deaths among women worldwide, including Thailand. In the present study, the differential mRNA expression of SVEP1, LPHN3, KLB, ITGA7, SEMA3G, TNS1 and MMP13 genes was examined in breast cancer using quantitative real-time
Mauricio Arcos-Burgos et al.
Attention deficit and hyperactivity disorders, 2(3), 139-147 (2011-03-25)
During the past 15 years, an impressive amount of genetic information has become available in the research field of psychiatry, particularly as it relates to attention-deficit/hyperactivity disorder (ADHD). However, the classical clinical approach to ADHD has minimally affected and not

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service