Skip to Content
Merck
All Photos(7)

Key Documents

HPA007187

Sigma-Aldrich

Anti-NELFE antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AL844853.23, Anti-NELF-E, Anti-Negative elongation factor E, Anti-RDBP, Anti-RNA-binding protein RD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

LLALKKQSSSSTTSQGGVKRSLSEQPVMDTATATEQAKQLVKSGAISAIKAETKNSGFKRSRTLEGKLKDPEKGPVPTFQPFQRSISADDDLQESSRRPQRKSLYESFVSSSDRLRELGPDG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RDBP(7936)

Immunogen

Negative elongation factor E recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NELFE (Negative elongation factor complex member E) is a major regulatory subunit of negative elongation factor (NELF) consisting of a conserved canonical βαββαβ RNA Recognition Motif (RRM). The RRM domain is mainly located at its β-sheet surface for identifying and binding to a wide variety of RNA sequences. It is mainly involved in the RNA Polymerase II (Pol II) transcript elongation. It directly binds to the nascent transcribed RNA for promoter-proximal RNA Pol II pausing of both HIV-1 and cellular genes. It has been suggested that NELFE may play a potential role in the prevention of hepatocellular carcinoma (HCC) cell metastasis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71152

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucy H Williams et al.
Molecular cell, 58(2), 311-322 (2015-03-17)
The remarkable capacity for pluripotency and self-renewal in embryonic stem cells (ESCs) requires a finely tuned transcriptional circuitry wherein the pathways and genes that initiate differentiation are suppressed, but poised to respond rapidly to developmental signals. To elucidate transcriptional control
Dependency of NELF-E-SLUG-KAT2B epigenetic axis in breast cancer carcinogenesis.
Zhang, et al.
Nature Communications, 14, 2439-2439 (2023)
Jampani Nageswara Rao et al.
Biochemistry, 47(12), 3756-3761 (2008-02-29)
The E subunit of the human heterotetrameric negative transcription elongation factor (NELF-E) contains a canonical betaalphabetabetaalphabeta RNA recognition motif (RRM) that binds to a wide variety of RNA sequences. These induce very similar conformational changes in the RRM as determined
Michihisa Iida et al.
Oncology reports, 28(2), 728-734 (2012-05-23)
Hepatocellular carcinoma (HCC) often exhibits a poor prognosis due to metastatic spread caused by portal vein invasion (PVI). In the present study, we attempted to identify a novel therapeutic target related to PVI of HCC. Based on pooled genomic data
Görkem Garipler et al.
Cell reports, 38(11), 110524-110524 (2022-03-17)
In pluripotent cells, a delicate activation-repression balance maintains pro-differentiation genes ready for rapid activation. The identity of transcription factors (TFs) that specifically repress pro-differentiation genes remains obscure. By targeting ∼1,700 TFs with CRISPR loss-of-function screen, we found that ZBTB11 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service