Skip to Content
Merck
All Photos(6)

Key Documents

HPA006104

Sigma-Aldrich

Anti-KAT6B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MOZ2, Anti-MYST4, Anti-Morf, Anti-ZC2HC6B, Anti-qkf, Anti-querkopf

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

KPVLRKAFQHQPGKKRQTEEEEGKDNHCFKNADPCRNNMNDDSSNLKEGSKDNPEPLKCKQVWPKGTKRGLSKWRQNKERKTGFKLNLYTPPETPMEPDEQVTVEEQKETSEGKTSPSPIRIEEEVKETGEAL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYST4(23522)

General description

Histone acetyltransferase MYST4 is an enzyme encoded by the MYST4 gene in humans and is mapped to chromosome band 10q22. It is referred to as qkf, MORF, MOZ2, GTPTS, MYST4, ZC2HC6B, querkopf and KAT6B. It is a 1781-residue protein and possesses similarity with MOZ (monocytic leukemia zinc finger protein). It is ubiquitously expressed in adult human tissues and has intrinsic histone acetyltransferase activity. It consists of histone acetyltransferase domain, a strong transcriptional repression domain at its N terminus and a strong activation domain at its C terminus.

Immunogen

Histone acetyltransferase MYST4 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KAT6B (K(lysine) acetyltransferase 6B) is considered as a novel histone acetyltransferase containing multiple functional domains and may be involved in both positive and negative regulation of transcription. This gene may play crucial role during early developmental processes. Mutation in this gene causes typical Say-Barber/Biesecker/Young-Simpson (SBBYS) syndrome and KAT6B-related disorders that includes genitopatellar syndrome (GPS).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70948

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hung-Chun Yu et al.
American journal of medical genetics. Part A, 164A(4), 950-957 (2014-01-25)
Blepharophimosis-ptosis-epicanthus inversus syndrome (BPES) is an autosomal dominant disorder caused by mutations in FOXL2. We identified an individual with BPES and additional phenotypic features who did not have a FOXL2 mutation. We used whole exome sequencing to identify a de
Margaret P Adam et al.
GeneReviews(?), 2012 Dec 13 (Updated 2013 10) (2013-01-10)
The diagnosis of
Katalin Szakszon et al.
American journal of medical genetics. Part A, 161A(4), 884-888 (2013-02-26)
The Say-Barber/Biesecker/Young-Simpson (SBBYS) type of the blepharophimosis-mental retardation syndrome group (Ohdo-like syndromes) is a multiple congenital malformation syndrome characterized by vertical narrowing and shortening of the palpebral fissures, ptosis, intellectual disability, hypothyroidism, hearing impairment, and dental anomalies. Mutations of the
N Champagne et al.
The Journal of biological chemistry, 274(40), 28528-28536 (1999-09-25)
We describe here the identification and functional characterization of a novel human histone acetyltransferase, termed MORF (monocytic leukemia zinc finger protein-related factor). MORF is a 1781-residue protein displaying significant sequence similarity to MOZ (monocytic leukemia zinc finger protein). MORF is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service