Skip to Content
Merck
All Photos(6)

Key Documents

HPA000781

Sigma-Aldrich

ANTI-ELFN2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ELFNO2, Anti-Extracellular leucine-rich repeat and fibronectin type III domain-containing protein 2, Anti-LRRC62, Anti-Leucine-rich repeat and fibronectin type-III domain-containing protein 6 precursor, Anti-Leucine-rich repeat-containing protein 62

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

GIHHLEVKPAYHCSEHRHSFPALYYEEGADSLSQRVSFLKPLTRSKRDSTYSQLSPRHYYSGYSSSPEYSSESTHKIWERFRPYKKHHREEVYMAAGHALRKKVQFAKDEDLHDILDYWK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ELFN2(114794)

Immunogen

Leucine-rich repeat and fibronectin type-III domain-containing protein 6 precursor (Extracellular leucine-rich repeat and fibronectin type III domain-containing protein 2) recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

Extracellular leucine-rich repeat and fibronectin type III domain containing 2 (ELFN2) is also known as LRRC62 and PPP1R29. The Leucine-rich repeats (LRRs) are highly versatile and evolvable protein-ligand interaction motifs. It is found in a large number of proteins with diverse functions that include innate immunity and nervous system development. The protein interacts with various regulatory proteins and may have a role in cell polarity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73532

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miklos Szoboszlay et al.
Scientific reports, 7(1), 15240-15240 (2017-11-12)
Nanoscale distribution of molecules within small subcellular compartments of neurons critically influences their functional roles. Although, numerous ways of analyzing the spatial arrangement of proteins have been described, a thorough comparison of their effectiveness is missing. Here we present an
Jackie Dolan et al.
BMC genomics, 8, 320-320 (2007-09-18)
Leucine-rich repeats (LRRs) are highly versatile and evolvable protein-ligand interaction motifs found in a large number of proteins with diverse functions, including innate immunity and nervous system development. Here we catalogue all of the extracellular LRR (eLRR) proteins in worms
Noemi Holderith et al.
Frontiers in synaptic neuroscience, 13, 773209-773209 (2022-03-01)
Selective distribution of proteins in presynaptic active zones (AZs) is a prerequisite for generating postsynaptic target cell type-specific differences in presynaptic vesicle release probability (Pv) and short-term plasticity, a characteristic feature of cortical pyramidal cells (PCs). In the hippocampus of
Annick Hendrickx et al.
Chemistry & biology, 16(4), 365-371 (2009-04-25)
The ubiquitous protein Ser/Thr phosphatase-1 (PP1) interacts with dozens of regulatory proteins that are structurally unrelated. However, most of them share a short, degenerate "RVxF"-type docking motif. Using a broad in silico screening based on a stringent definition of the
Tevye Jason Stachniak et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 39(23), 4461-4474 (2019-04-04)
Excitatory synapses onto somatostatin (SOM) interneurons show robust short-term facilitation. This hallmark feature of SOM interneurons arises from a low initial release probability that regulates the recruitment of interneurons in response to trains of action potentials. Previous work has shown

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service