Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5441-22

HiTrap® Capto S

Cytiva 17-5441-22, pack of 5 × 1 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NA.56

ligand

sulfonate group

description

Ion Exchanger Type (value)

packaging

pack of 5 × 1 mL

manufacturer/tradename

Cytiva 17-5441-22

parameter

<4 mL/min flow rate
42 psi

bed size

7 mm × 25 mm

bed volume

1 mL

column I.D.

7 mm

matrix

highly cross-linked agarose with dextran surface extender

average diameter

90 μm (d50v)

cleaning

3-14

working range

4-13

suitability

suitable for bioprocess medium

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

HiTrap® Capto S is a strong cation exchanger prepacked with BioProcess Capto media for screening and small-scale protein purifications using ion exchange chromatography (IEX).

Application

Capto S has a strong sulfoethyl cation exchanger coupled to a chemically modified, high-fLow Agarose matrix. The modified Agarose matrix provides greater particle rigidity without compromising pore size, which allows more flexibility in process-scale applications. High dynamic binding capacity is achieved with a Dextran surface extender that coats the Agarose matrix.

Features and Benefits

  • High-throughput medium for capture and intermediate purification, excellent choice for process-scale purification.
  • The combined properties of high binding capacity, high flow rate, and Low back pressure reduces process cycle times and improves productivity at process scale.
  • Rigid Agarose matrix enables high bed heights and purification of viscous samples at high flow rates.
  • Strong sulfoethyl(S) cation exchanger.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (0.2 M sodium acetate in 20% ethanol)
Store at 4 to 30 °C (20% Ethanol + 0.2 M Sodium Acetate)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Other Notes

The pressure over the packed bed varies depending on a range of parameters such as the characteristics of the chromatography medium and the column tubing used.

Legal Information

Capto is a trademark of Cytiva
HiTrap is a registered trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protocols

This page covers detailed information on cleaning procedures and recommended flow for column cleaning.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service