GE10600117
Amersham™ Protran® Premium Western blotting membrane sandwich, nitrocellulose
pore size 0.45 μm, sheet W × L 80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers, pkg of 10 ea
About This Item
Recommended Products
packaging
pkg of 10 ea
manufacturer/tradename
Cytiva 10600117
sheet W × L
80 mm × 90 mm , preassembled with 2 x 3MM chr filter papers
pore size
0.45 μm pore size
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- Precut Amersham Protran Premium 0.45 µm nitrocellulose membranes.
Storage and Stability
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
11 - Combustible Solids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service