EP951040021
Eppendorf® Microplate 96/F-PP
sterile, F-bottom, clear Wells, white border, pkg of 80 plates (5 bags x 16 plates)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122107
material:
colorless wells
flat bottom wells
flat bottom wells
size:
96 well
sterility:
sterile
feature:
F-bottom
Recommended Products
material
colorless wells
flat bottom wells
sterility
sterile
feature
F-bottom
packaging
pkg of 80 plates (5 bags x 16 plates)
manufacturer/tradename
Eppendorf® 951040021 (North America)
technique(s)
PCR: suitable
size
96 well
color
clear Wells
white border
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service