Key Documents
EP951031208
Eppendorf® Deepwell plates, DNA LoBind, 384 wells
white plate, conical bottom, colorless wells, capacity 200 μL, pack of 40 ea (5 bags × 8 plates)
About This Item
UNSPSC Code:
41106300
material
clear wells
colorless wells
conical bottom
polypropylene
white plate
sterility
non-sterile
feature
lid: no
packaging
pack of 40 ea (5 bags × 8 plates)
manufacturer/tradename
Eppendorf® 951031208
capacity
200 μL
well volume
250 μL
color
white border
suitability
suitable for PCR
suitable for molecular biology
binding type
low binding surface
Related Categories
General description
Deepwell Plate 384/200µL, DNA LoBind, wells clear, 200 µL, LoBind, PCR clean, white, 40 plates (5 bags × 8 plates)
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing
Features and Benefits
  • Eppendorf LoBind material ensures optimized sample recovery for improved assay results
  • Free of surface coating (e.g. silicone) to minimize the risk of sample interference
  • Lot-certified PCR clean purity grade: free of human DNA, DNase, RNase and PCR inhibitors
  • Available in tube, microplate, and deepwell plate formats for easy-up scaling
  • High-contrast Unique OptiTrack® matrix: up to 30 % faster sample identification and fewer pipetting errors
  • RecoverMax® well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
  • Raised well rims and a smooth surface ensure reliable sealing
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
OptiTrack is a registered trademark of Eppendorf AG
RecoverMax is a registered trademark of Eppendorf AG
Visit the Document Library
Peptidomics of the agriculturally damaging larval stage of the cabbage root fly Delia radicum (Diptera: Anthomyiidae)
Judith Zoephel
PLoS ONE, 7, 7-7 (2012)
Human tear peptide/protein profiling study of ocular surface diseases by SPE-MALDI-TOF mass spectrometry analyses
Nerea Gonzalez a 1
EuPA open proteomics, 3, 206-215 (2014)
Peptidomics of the agriculturally damaging larval stage of the cabbage root fly Delia radicum (Diptera: Anthomyiidae)
Judith Zoephel
PLoS ONE, 7, 7-7 (2012)
Affinity Purification of Yeast Protein-interacting Metabolites for ESI-MS Analysis
Xiyan Li
Bio-protocol, 1 (2011)
E I Trilisky et al.
Journal of chromatography. A, 1142(1), 2-12 (2007-01-24)
Purified viruses are used in gene therapy and vaccine production. Ion-exchange chromatography (IEC) is the most common method for large-scale downstream purification of viruses and proteins. Published IEC protocols provide details for specific separations but not general methods for selecting