EP5702733008
Adapter for Eppendorf® A-4-38 Rotor
holds 1 Eppendorf Tube® 5.0 mL, for use with 100 mL round buckets, pack of 2 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
flat bottom
feature
holds 1 Eppendorf Tube® 5.0 mL,
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702733008 (Global)
parameter
2.943 × g max. RCF
technique(s)
centrifugation: suitable
diam.
60 mm
compatibility
for use with 100 mL round buckets
Looking for similar products? Visit Product Comparison Guide
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf Tube is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service