EP022639911
Rotor for Eppendorf® 5804/5804R & 5810/5810R Centrifuges
FA-45-48-11, aerosol-tight
All Photos(1)
About This Item
UNSPSC Code:
41103909
Recommended Products
material
aluminum
feature
RCF 19,083 × g
aerosol-tight
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 022639911
compatibility
5804/5804R & 5810/5810R
FA-45-48-11
Related Categories
General description
Rotor FA-45-48-11, aerosol-tight
1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information. 2) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain.
- 48 places for 1.5/2.0 mL tubes
- Max. speed: 19,083 × g (max. 13,000 rpm)1)
- Eppendorf QuickLock® rotor lid for fast lid locking
- Aerosol-tight for safe centrifugation of hazardous samples2)
- Anodized aluminum for high chemical resistance
- Rotor and lid are autoclavable
1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information. 2) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain.
Features and Benefits
48 places for 1.5/2.0 mL tubes
Max. speed: 19,083 × g (max. 13,000 rpm)1)
Eppendorf QuickLock® rotor lid for fast lid locking
Aerosol-tight for safe centrifugation of hazardous samples2)
Anodized aluminum for high chemical resistance
Rotor and lid are autoclavable
1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information. 2) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain.
Max. speed: 19,083 × g (max. 13,000 rpm)1)
Eppendorf QuickLock® rotor lid for fast lid locking
Aerosol-tight for safe centrifugation of hazardous samples2)
Anodized aluminum for high chemical resistance
Rotor and lid are autoclavable
1) Please note that a software upgrade may be necessary for older centrifuges when using this rotor. Please contact your local Eppendorf organization or dealer for further information. 2) Aerosol-tightness tested by Public Health England, Porton Down, Great Britain.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf QuickLock is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)