EP022492098
Eppendorf® epT.I.P.S.® standard
volume 0.5-10 mL, L 165 mm, turquoise, colorless tip, pkg of 200 ea (2 bags × 100 tips)
Synonym(s):
pipette tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
colorless tip
polypropylene
turquoise
sterility
non-sterile
packaging
pkg of 200 ea (2 bags × 100 tips)
manufacturer/tradename
Eppendorf® 022492098
L
165 mm
volume
0.5-10 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
epT.I.P.S.® Standard, Eppendorf Quality, 0.5 – 10 mL, 165 mm, turquoise, colorless tips, 200 tips (2 bags × 100 tips)
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade
- epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Features and Benefits
- Original, high-quality Eppendorf pipette tips packaged in resealable bags
- Available in sizes from 10 µL to 10 mL in Eppendorf Quality purity grade
- epT.I.P.S. Standard 200 µL, 300 µL and 1,000 µL tips are also available color-coded yellow and blue
- epT.I.P.S. Standard pipette tips can be autoclaved at 121 °C in 20 min.
- Fine retracted or extended tips simplify work with small volumes
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
epT.I.P.S. is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service