Skip to Content
Merck
All Photos(1)

Documents

EP0030119819

Eppendorf® Tube Rack for 1.5 and 2.0 mL Tubes

Holds 36 x 1.5-2 mL tubes, 3 rows of 12 wells each, pkg of 2 pieces, white, autoclavable

Synonym(s):

Eppendorf rack

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122800
NACRES:
NB.12

material

polypropylene

feature

autoclavable
3 rows of 12 wells each
Holds 36 x 1.5-2 mL tubes

packaging

pkg of 2 pieces

manufacturer/tradename

Eppendorf® 30119819

color

white

General description

Eppendorf® Tube Rack for 1.5 and 2.0 mL Tubes are suitable for all laboratory protocols that require efficient and reliable processing, transport, and short-term storage of sample vessels in benchtop racks.

Features and Benefits

  • The design of the racks allows for both stackings within one rack format and between all rack variants, which helps to save valuable laboratory space when the racks are not in use.
  • The centrifuge racks are equipped with non-slip silicone feet and a tilt-proof footprint design to ensure safe handling and stability.
  • The wavelike design of the racks arranges tube rows at different heights, making it easy to access and identify the reaction vessels.
  • The racks are constructed using polypropylene (PP), providing them with high resistance to chemicals and physical stress.
  • The dimensions of the racks are optimized for space-saving, maintaining a perfect tube position-to-footprint arrangement without compromising tube handling capabilities.
  • The racks offer high flexibility as they can accommodate both 1.5 mL and 2.0 mL tubes. Each tube position is numbered, facilitating quick and easy orientation and ensuring error-free handling of the tubes.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service