Skip to Content
Merck
All Photos(1)

Key Documents

EHU143641

Sigma-Aldrich

MISSION® esiRNA

targeting human FLT4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GACACCTGGACCGAGTTTGTGGAGGGAAAGAATAAGACTGTGAGCAAGCTGGTGATCCAGAATGCCAACGTGTCTGCCATGTACAAGTGTGTGGTCTCCAACAAGGTGGGCCAGGATGAGCGGCTCATCTACTTCTATGTGACCACCATCCCCGACGGCTTCACCATCGAATCCAAGCCATCCGAGGAGCTACTAGAGGGCCAGCCGGTGCTCCTGAGCTGCCAAGCCGACAGCTACAAGTACGAGCATCTGCGCTGGTACCGCCTCAACCTGTCCACGCTGCACGATGCGCACGGGAACCCGCTTCTGCTCGACTGCAAGAACGTGCATCTGTTCGCCACCCCTCTGGCCGCCAGCCTGGAGGAGGTGGCACCTGGGGCGCGCCACGCCACGCTCAGCCTGAGTATCCCCCGCGTCGCGCCCGAGCACGAGGGCCACTATGTGTGCGAAGTGCAAGACCGGCGCAGCCATGACAAGCACTGCCACAAGAA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joo-Hee Park et al.
PloS one, 9(9), e109055-e109055 (2014-10-01)
MAZ51 is an indolinone-based molecule originally synthesized as a selective inhibitor of vascular endothelial growth factor receptor (VEGFR)-3 tyrosine kinase. This study shows that exposure of two glioma cell lines, rat C6 and human U251MG, to MAZ51 caused dramatic shape
Yan Zhang et al.
Nature communications, 9(1), 1296-1296 (2018-04-05)
Incomplete delivery to the target cells is an obstacle for successful gene therapy approaches. Here we show unexpected effects of incomplete targeting, by demonstrating how heterogeneous inhibition of a growth promoting signaling pathway promotes tissue hyperplasia. We studied the function
Sungwoon Lee et al.
The Journal of clinical investigation, 127(2), 457-471 (2016-12-20)
Controlled angiogenesis and lymphangiogenesis are essential for tissue development, function, and repair. However, aberrant neovascularization is an essential pathogenic mechanism in many human diseases, including diseases involving tumor growth and survival. Here, we have demonstrated that mice deficient in C-type
Raj Kumar et al.
Cell death & disease, 11(5), 325-325 (2020-05-10)
Pathological retinal neovascularization is the most common cause of vision loss. PKCθ has been shown to play a role in type 2 diabetes, which is linked to retinal neovascularization. Based on these clues, we have studied the role of PKCθ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service