Skip to Content
Merck
All Photos(1)

Key Documents

EHU037241

Sigma-Aldrich

MISSION® esiRNA

targeting human SMAD3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GGCTTTGAGGCTGTCTACCAGTTGACCCGAATGTGCACCATCCGCATGAGCTTCGTCAAAGGCTGGGGAGCGGAGTACAGGAGACAGACTGTGACCAGTACCCCCTGCTGGATTGAGCTGCACCTGAATGGGCCTTTGCAGTGGCTTGACAAGGTCCTCACCCAGATGGGCTCCCCAAGCATCCGCTGTTCCAGTGTGTCTTAGAGACATCAAGTATGGTAGGGGAGGGCAGGCTTGGGGAAAATGGCCATGCAGGAGGTGGAGAAAATTGGAACTCTACTCAACCCATTGTTGTCAAGGAAGAAGAAATCTTTCTCCCTCAACTGAAGGGGTGCACCCACCTGTTTTCTGAAACACACGAGCAAACCCAGAGGTGGATGTTATGAACAGCTGTGTCTGCCAAACACATTTACCCTTTGGCCCCACTTTGAAGGGCAAGAAATGGCGTCTGCTCTGGTGGCTTAAGTGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Muppala et al.
Oncogene, 36(36), 5189-5198 (2017-05-10)
TGF-β is a multifunctional cytokine affecting many cell types and implicated in tissue remodeling processes. Due to its many functions and cell-specific effects, the consequences of TGF-β signaling are process-and stage-dependent, and it is not uncommon that TGF-β exerts distinct
Nao Hiwatashi et al.
The Laryngoscope, 127(9), E308-E316 (2017-05-26)
Recent reports highlight the efficacy of small interfering RNA (siRNA) targeting SMAD3 to regulate transforming growth factor β (TGF-β)-mediated fibroplasia in vocal fold fibroblasts. The current study sought to investigate SMAD3 expression during wound healing in vivo and quantify the
Zheng Jiang et al.
Journal of biochemistry, 165(4), 317-322 (2018-12-12)
Radiotherapy is the major treatment modality for malignant glioma. However, the treatment response of radiotherapy is suboptimal due to resistance. Here we aimed to explore the effect and mechanism of Mothers against decapentaplegic homologue (SMAD3) silencing in sensitizing malignant glioma
Ayesha Ghayur et al.
American journal of physiology. Renal physiology, 317(1), F152-F162 (2019-05-30)
Glomerulonephritis (GN) is a common cause of end-stage kidney disease and is characterized by glomerular inflammation, hematuria, proteinuria, and progressive renal dysfunction. Transforming growth factor (TGF)-β is involved in glomerulosclerosis and interstitial fibrosis. TGF-β activates multiple signaling pathways, including the
Tianli Cheng et al.
International journal of oncology, 45(5), 1977-1988 (2014-09-02)
Altered expression of miRNAs contributes to development and progression of non-small cell lung cancer (NSCLC), while transforming growth factor-β (TGF-β) promotes NSCLC cell epithelial-mesenchymal transition. This study aimed to investigate the effects of TGF-β-induced miR‑143 expression in regulation of NSCLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service