Skip to Content
Merck
All Photos(1)

Documents

D8190

Sigma-Aldrich

3′,4′-Dichlorobenzamil hydrochloride

>98% (HPLC)

Synonym(s):

L-594,881

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H12N7OCl3 · HCl
CAS Number:
Molecular Weight:
425.10
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>98% (HPLC)

form

solid

color

yellow

solubility

DMSO: soluble 30 mg/mL
H2O: insoluble

storage temp.

2-8°C

SMILES string

NC1=NC(N)=C(Cl)N=C1C(NC(NCC2=CC(Cl)=C(Cl)C=C2)=N)=O.Cl

InChI

1S/C13H12Cl3N7O.ClH/c14-6-2-1-5(3-7(6)15)4-20-13(19)23-12(24)8-10(17)22-11(18)9(16)21-8;/h1-3H,4H2,(H4,17,18,22)(H3,19,20,23,24);1H

InChI key

LPHAOBPPRPXQLQ-UHFFFAOYSA-N

Biochem/physiol Actions

Inhibits Na+/Ca2+ exchanger, Na+ transport and sarcoplasmic reticulum Ca2+ release channels.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manu Hegde et al.
The Journal of pharmacology and experimental therapeutics, 310(1), 67-74 (2004-03-11)
Previously, we demonstrated that malignant glioma cell lines have increased intracellular pH (pHi) as a result of increased activities of the type I sodium/hydrogen exchanger (NHE1). This alkalotic pHi of 7.2 to 7.4 is favorable for augmented glycolysis, DNA synthesis
Toru Oda et al.
Journal of pharmacological sciences, 116(1), 128-131 (2011-04-28)
To elucidate the role of Na(+)/Ca(2+) exchanger (NCX) in neurite outgrowth, we investigated the effects of NCX inhibitors on neurite outgrowth in PC12 cells. KB-R7943 and 3',4'-dichlorobenzamil, NCX inhibitors, inhibited the neurite outgrowth caused by nerve growth factor (NGF). NCX
A S Arbab et al.
Nuclear medicine communications, 24(2), 155-166 (2003-01-28)
Our aim was to determine whether the Ca2+ ion or cell membrane Ca2+ and Na+/Ca2+ ion transport systems are involved in maintaining the cell-associated activity of technetium-99m-hexakis-methoxy-isobutyl-isonitrile (99mTc-MIBI) and technetium-99m-ethylene-bis[bis(2-ethoxyethyl)phosphin] (99mTc-tetrofosmin) in tumour cell lines. The cell-associated activities of 99mTc-MIBI
Takahiro Horinouchi et al.
European journal of pharmacology, 566(1-3), 28-33 (2007-04-21)
Endothelin ET(A) receptor couples to Gq/11 protein that transduces a variety of receptor signals to modulate diverse cellular responses including Ca2+ mobilization. Stimulation of endothelin ETA receptor with endothelin-1 is generally believed to induce an increase in intracellular Ca2+ concentration
Abhishek Gulati et al.
The Journal of pharmacy and pharmacology, 58(8), 1145-1151 (2006-07-29)
Elevation of intrasperm Ca(2+) is reported to influence viability of ejaculated spermatozoa. Human spermatozoa possess inositol triphosphate (IP(3))-sensitive Ca(2+) stores, which can be targeted for increasing intrasperm Ca(2+) level. The influence of agents affecting Ca(2+) homeostasis has been investigated. Miconazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service